Assay Details
Gene Symbol: Tsn | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_004836025.3 | XP_004836082.1 | 2 - 3 | 332 | 63 | |
XM_004836026.3 | XP_004836083.1 | 2 - 3 | 344 | 63 |

More Information
Set Membership: |
![]() ![]() ![]() |
