Assay Details
Gene Symbol: Mcrip1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_004860958.3 | XP_004861015.1 | 3 - 4 | 235 | 73 | |
XM_021241184.1 | XP_021096843.1 | 3 - 4 | 438 | 73 |

More Information
Set Membership: |
![]() ![]() ![]() |
