Assay Details
Gene Symbol: Mcrip1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_004860958.3 | XP_004861015.1 | 4 - 5 | 338 | 71 | |
XM_021241184.1 | XP_021096843.1 | 4 - 5 | 541 | 71 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
