Assay Details
| Gene Symbol: Slc26a1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004847458.3 | XP_004847515.1 | 2 - 3 | 1406 | 57 | |
| XM_004847459.3 | XP_004847516.1 | 2 - 3 | 771 | 57 | ||
| XM_004847460.3 | XP_004847517.1 | 2 - 3 | 618 | 57 | ||
| XM_004847461.3 | XP_004847518.1 | 2 - 3 | 856 | 57 | ||
More Information
Set Membership: |
|

Slc26a1
Genomic Map