Assay Details
| Gene Symbol: Nmnat1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004863631.3 | XP_004863688.1 | 2 - 3 | 264 | 67 | |
| XM_004863632.3 | XP_004863689.1 | 2 - 3 | 264 | 67 | ||
More Information
Set Membership: |
|

Nmnat1
Genomic Map