Assay Details
| Gene Symbol: Cct5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004845180.3 | XP_004845237.1 | 5 - 6 | 823 | 65 | |
| XM_021258606.1 | XP_021114265.1 | 5 - 6 | 741 | 65 | ||
More Information
Set Membership: |
|

Cct5
Genomic Map