Assay Details
Gene Symbol: Spg21 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_004855638.3 | XP_004855695.1 | 8 - 9 | 1050 | 66 | |
XM_021237071.1 | XP_021092730.1 | 9 - 10 | 1464 | 66 | ||
XM_021237072.1 | XP_021092731.1 | 8 - 9 | 999 | 66 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
