Assay Details
| Gene Symbol: Aes | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004865825.3 | XP_004865882.1 | 6 - 7 | 446 | 52 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

Aes| Gene Symbol: Aes | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004865825.3 | XP_004865882.1 | 6 - 7 | 446 | 52 | |
Set Membership: |
|