Assay Details
| Gene Symbol: Tmem205 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004865575.3 | XP_004865632.1 | 3 - 4 | 462 | 64 | |
| XM_004865576.3 | XP_004865633.1 | 3 - 4 | 457 | 64 | ||
| XM_021244981.1 | XP_021100640.1 | 3 - 4 | 298 | 64 | ||
More Information
Set Membership: |
|

Tmem205
Genomic Map