Assay Details
| Gene Symbol: Atp5pd | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004860630.3 | XP_004860687.2 | 5 - 6 | 517 | 81 | |
| XM_004860632.3 | XP_004860689.2 | 4 - 5 | 430 | 81 | ||
More Information
Set Membership: |
|

Atp5pd
Genomic Map