Assay Details
| Gene Symbol: Atp5f1a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004855084.3 | XP_004855141.1 | 4 - 5 | 597 | 64 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

Atp5f1a| Gene Symbol: Atp5f1a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004855084.3 | XP_004855141.1 | 4 - 5 | 597 | 64 | |
Set Membership: |
|