Assay Details
| Gene Symbol: S100a1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004871435.3 | XP_004871492.1 | 1 - 2 | 209 | 67 | |
More Information
Additional Information:
This assay is in the 5'UTR region of the XM_004871435.3 transcripts.
Set Membership: |
|

S100a1
Genomic Map