Assay Details
Gene Symbol: Ccdc127 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_004845100.3 | XP_004845157.1 | 2 - 3 | 316 | 77 | |
XM_021258556.1 | XP_021114215.1 | 3 - 4 | 297 | 77 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
