Assay Details
| Gene Symbol: Apoc2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004873144.3 | XP_004873201.1 | 3 - 4 | 426 | 62 | |
| XM_004873146.3 | XP_004873203.1 | 3 - 4 | 357 | 62 | ||
More Information
Set Membership: |
|

Apoc2
Genomic Map