Assay Details
| Gene Symbol: Hsd3b7 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004856210.3 | XP_004856267.1 | 6 - 7 | 961 | 61 | |
| GenBank mRNA | KM486724.1 | - | 6 - 7 | 863 | 61 | |
More Information
Set Membership: |
|

Hsd3b7
Genomic Map