Assay Details
| Gene Symbol: Fen1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_013069142.2 | XP_012924596.1 | 2 | 1139 | 125 | |
| XM_013069143.2 | XP_012924597.1 | 2 | 1110 | 125 | ||
| XM_021251748.1 | XP_021107407.1 | 3 | 1306 | 125 | ||
| XM_021251749.1 | XP_021107408.1 | 4 | 1415 | 125 | ||
More Information
Set Membership: |
|

Fen1
Genomic Map