Assay Details
| Gene Symbol: Fam174a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004841731.2 | XP_004841788.1 | 1 - 2 | 649 | 69 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

Fam174a| Gene Symbol: Fam174a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004841731.2 | XP_004841788.1 | 1 - 2 | 649 | 69 | |
Set Membership: |
|