Assay Details
| Gene Symbol: Rpl10a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004846711.3 | XP_004846768.1 | 4 - 5 | 467 | 80 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

Rpl10a| Gene Symbol: Rpl10a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004846711.3 | XP_004846768.1 | 4 - 5 | 467 | 80 | |
Set Membership: |
|