Assay Details
Gene Symbol: LOC101715817 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_004851993.3 | XP_004852050.1 | 12 - 13 | 2144 | 69 | |
XM_004851995.3 | XP_004852052.2 | 14 - 15 | 2852 | 69 |

More Information
Set Membership: |
![]() ![]() |
Panther Classification:
