Assay Details
| Gene Symbol: Agt | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004854676.2 | XP_004854733.1 | 4 - 5 | 1476 | 69 | |
| XM_021266424.1 | XP_021122083.1 | 4 - 5 | 1487 | 69 | ||
More Information
Set Membership: |
|

Agt
Genomic Map