Assay Details
Gene Symbol: Itprip | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_004866507.2 | XP_004866564.1 | 2 - 3 | 1225 | 88 | |
XM_004866509.2 | XP_004866566.1 | 2 - 3 | 539 | 88 | ||
XM_021245553.1 | XP_021101212.1 | 3 - 4 | 778 | 88 | ||
XM_021245554.1 | XP_021101213.1 | 3 - 4 | 2017 | 88 | ||
XM_021245555.1 | XP_021101214.1 | 2 - 3 | 633 | 88 | ||
XM_021245556.1 | XP_021101215.1 | 2 - 3 | 1872 | 88 | ||
XM_021245557.1 | XP_021101216.1 | 2 - 3 | 2175 | 88 |

More Information
Additional Information:
This assay is in the 5'UTR region of the XM_004866507.2, XM_004866509.2, XM_021245553.1, XM_021245554.1, XM_021245555.1, XM_021245556.1, XM_021245557.1 transcripts.
Set Membership: |
![]() ![]() ![]() ![]() |
