Assay Details
Gene Symbol: Cmc4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_004875178.3 | XP_004875235.1 | 2 - 3 | 261 | 73 | |
XM_004875179.2 | XP_004875236.1 | 2 - 3 | 257 | 73 | ||
XM_004875180.3 | XP_004875237.1 | 2 - 3 | 249 | 73 | ||
XM_004875181.2 | XP_004875238.1 | 2 - 3 | 259 | 73 | ||
XM_013069451.2 | XP_012924905.1 | 2 - 3 | 209 | 73 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
