Assay Details
Gene Symbol: Tedc1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_013066060.2 | XP_012921514.1 | 4 - 5 | 788 | 75 | |
XM_013066061.2 | XP_012921515.1 | 4 - 5 | 788 | 75 | ||
XM_013066062.2 | XP_012921516.1 | 4 - 5 | 788 | 75 | ||
XM_021243258.1 | XP_021098917.1 | 4 - 5 | 788 | 75 | ||
XM_021243259.1 | XP_021098918.1 | 4 - 5 | 1197 | 75 | ||
XM_021243261.1 | XP_021098920.1 | 4 - 5 | 509 | 75 | ||
XM_021243266.1 | XP_021098925.1 | 4 - 5 | 788 | 75 |

More Information
Set Membership: |
![]() ![]() ![]() |
