Assay Details
Gene Symbol: Nif3l1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_013068115.2 | XP_012923569.1 | 5 - 6 | 1015 | 55 | |
XM_013068116.2 | XP_012923570.1 | 5 - 6 | 1876 | 55 | ||
XM_013068117.2 | XP_012923571.1 | 5 - 6 | 981 | 55 | ||
XM_013068118.2 | XP_012923572.1 | 5 - 6 | 971 | 55 | ||
XM_021249756.1 | XP_021105415.1 | 5 - 6 | 1200 | 55 | ||
XM_021249757.1 | XP_021105416.1 | 6 - 7 | 1394 | 55 | ||
XM_021249758.1 | XP_021105417.1 | 6 - 7 | 1723 | 55 | ||
XM_021249759.1 | XP_021105418.1 | 5 - 6 | 1137 | 55 | ||
XM_021249760.1 | XP_021105419.1 | 6 - 7 | 2257 | 55 | ||
XM_021249762.1 | XP_021105421.1 | 5 - 6 | 906 | 55 |

More Information
Set Membership: |
![]() ![]() ![]() |
