Assay Details
Gene Symbol: Kdelc2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_021237863.1 | XP_021093522.1 | 5 - 6 | 1396 | 67 | |
XM_021237864.1 | XP_021093523.1 | 5 - 6 | 967 | 67 |

More Information
Set Membership: |
![]() ![]() ![]() |
