Assay Details
| Gene Symbol: LOC101713466 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_021248315.1 | XP_021103974.1 | 5 - 6 | 3356 | 99 | |
| XM_021248316.1 | XP_021103975.1 | 3 - 4 | 335 | 99 | ||
| XM_021248317.1 | XP_021103976.1 | 3 - 4 | 805 | 99 | ||
More Information
Set Membership: |
|

LOC101713466
Genomic Map