Assay Details
Gene Symbol: LOC101703656 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_004837769.3 | XP_004837826.1 | 2 - 3 | 215 | 95 | |
XM_013069270.2 | XP_012924724.1 | 2 - 3 | 422 | 95 | ||
XM_013069272.2 | XP_012924726.1 | 2 - 3 | 504 | 95 | ||
XM_021250448.1 | XP_021106107.1 | 10 - 11 | 2881 | 95 | ||
XM_021250452.1 | XP_021106111.1 | 10 - 11 | 2880 | 95 |

More Information
Set Membership: |
![]() ![]() ![]() |
Panther Classification:
