Assay Details
Gene Symbol: LOC101703656 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_004837769.3 | XP_004837826.1 | 5 - 6 | 383 | 65 | |
XM_004837771.2 | XP_004837828.1 | 4 - 5 | 421 | 65 | ||
XM_004837772.2 | XP_004837829.1 | 4 - 5 | 334 | 65 | ||
XM_004837778.3 | XP_004837835.1 | 4 - 5 | 271 | 65 | ||
XM_013069270.2 | XP_012924724.1 | 5 - 6 | 590 | 65 | ||
XM_013069272.2 | XP_012924726.1 | 5 - 6 | 672 | 65 | ||
XM_013069280.2 | XP_012924734.1 | 4 - 5 | 339 | 65 | ||
XM_013069294.2 | XP_012924748.1 | 3 - 4 | 406 | 65 | ||
XM_013069297.2 | XP_012924751.1 | 3 - 4 | 319 | 65 | ||
XM_013069315.2 | XP_012924769.1 | 4 - 5 | 414 | 65 | ||
XM_021250448.1 | XP_021106107.1 | 13 - 14 | 3049 | 65 | ||
XM_021250456.1 | XP_021106115.1 | 4 - 5 | 901 | 65 | ||
XM_021250459.1 | XP_021106118.1 | 3 - 4 | 886 | 65 | ||
XM_021250488.1 | XP_021106147.1 | 3 - 4 | 324 | 65 | ||
XM_021250490.1 | XP_021106149.1 | 3 - 4 | 399 | 65 | ||
XM_021250524.1 | XP_021106183.1 | 4 - 5 | 254 | 65 | ||
XM_021250534.1 | XP_021106193.1 | 3 - 4 | 255 | 65 | ||
XM_021250542.1 | XP_021106201.1 | 3 - 4 | 239 | 65 |

More Information
Set Membership: |
![]() ![]() ![]() |
Panther Classification:
