Assay Details
Gene Symbol: Hs1bp3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_004839063.3 | XP_004839120.1 | 2 - 3 | 229 | 86 | |
XM_004839065.2 | XP_004839122.1 | 2 - 3 | 2908 | 86 | ||
XM_004839066.3 | XP_004839123.1 | 2 - 3 | 295 | 86 | ||
XM_004839067.3 | XP_004839124.1 | 2 - 3 | 229 | 86 | ||
XM_021253535.1 | XP_021109194.1 | 2 - 3 | 229 | 86 |

More Information
Set Membership: |
![]() ![]() ![]() |
