Assay Details
| Gene Symbol: Pdxk | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004842500.3 | XP_004842557.1 | 6 - 7 | 569 | 63 | |
| XM_013071376.2 | XP_012926830.1 | 6 - 7 | 569 | 63 | ||
| XM_021256382.1 | XP_021112041.1 | 5 - 6 | 464 | 63 | ||
More Information
Set Membership: |
|

Pdxk
Genomic Map