Assay Details
| Gene Symbol: Bri3bp | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004844101.2 | XP_004844158.1 | 1 - 2 | 303 | 66 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

Bri3bp| Gene Symbol: Bri3bp | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004844101.2 | XP_004844158.1 | 1 - 2 | 303 | 66 | |
Set Membership: |
|