Assay Details
Gene Symbol: Dcx | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_004861426.2 | XP_004861483.1 | 6 - 7 | 1139 | 86 | |
XM_004861427.3 | XP_004861484.1 | 6 - 7 | 1130 | 86 | ||
XM_004861428.2 | XP_004861485.1 | 6 - 7 | 1145 | 86 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
