Assay Details
Gene Symbol: Dffa | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_004863602.2 | XP_004863659.1 | 2 - 3 | 337 | 63 | |
XM_004863603.2 | XP_004863660.1 | 2 - 3 | 336 | 63 |

More Information
Set Membership: |
![]() ![]() ![]() |
