Assay Details
Gene Symbol: Shb | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_004864981.3 | XP_004865038.2 | 3 - 4 | 1656 | 77 | |
XM_021244508.1 | XP_021100167.1 | 4 - 5 | 1713 | 77 | ||
XM_021244509.1 | XP_021100168.1 | 4 - 5 | 1717 | 77 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
