Assay Details
| Gene Symbol: Npff | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004872754.3 | XP_004872811.1 | 2 - 3 | 1851 | 73 | |
| XM_004872755.3 | XP_004872812.1 | 2 - 3 | 825 | 73 | ||
More Information
Set Membership: |
|

Npff
Genomic Map