Assay Details
| Gene Symbol: Sec22a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004873618.3 | XP_004873675.1 | 3 - 4 | 486 | 76 | |
| XM_021251272.1 | XP_021106931.1 | 3 - 4 | 486 | 76 | ||
| XM_021251273.1 | XP_021106932.1 | 3 - 4 | 486 | 76 | ||
| XM_021251274.1 | XP_021106933.1 | 3 - 4 | 486 | 76 | ||
More Information
Set Membership: |
|

Sec22a
Genomic Map