Assay Details
| Gene Symbol: Shroom2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_013066472.2 | XP_012921926.1 | 3 - 4 | 579 | 54 | |
| XM_021246344.1 | XP_021102003.1 | 3 - 4 | 579 | 54 | ||
| XM_021246345.1 | XP_021102004.1 | 3 - 4 | 579 | 54 | ||
| XM_021246346.1 | XP_021102005.1 | 3 - 4 | 579 | 54 | ||
| XM_021246347.1 | XP_021102006.1 | 3 - 4 | 579 | 54 | ||
| XM_021246348.1 | XP_021102007.1 | 3 - 4 | 579 | 54 | ||
| XM_021246349.1 | XP_021102008.1 | 3 - 4 | 579 | 54 | ||
| XM_021246350.1 | XP_021102009.1 | 3 - 4 | 579 | 54 | ||
More Information
Set Membership: |
|

Shroom2
Genomic Map