Assay Details
| Gene Symbol: LOC101698217 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_013073201.2 | XP_012928655.2 | 4 - 5 | 466 | 83 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

LOC101698217| Gene Symbol: LOC101698217 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_013073201.2 | XP_012928655.2 | 4 - 5 | 466 | 83 | |
Set Membership: |
|