Assay Details
| Gene Symbol: Zpbp2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_021240076.1 | XP_021095735.1 | 5 - 6 | 831 | 66 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

Zpbp2| Gene Symbol: Zpbp2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_021240076.1 | XP_021095735.1 | 5 - 6 | 831 | 66 | |
Set Membership: |
|