Assay Details
Gene Symbol: Ccdc160 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_021240619.1 | XP_021096278.1 | 2 - 3 | 348 | 73 |

More Information
Additional Information:
This assay is in the 5'UTR region of the XM_021240619.1 transcripts.
Set Membership: |
![]() ![]() ![]() |
