Assay Details
Gene Symbol: Dcun1d1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_004834660.3 | XP_004834717.1 | 6 - 7 | 839 | 83 | |
XM_004834661.3 | XP_004834718.1 | 6 - 7 | 838 | 83 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
