Assay Details
| Gene Symbol: LOC101696584 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004843079.2 | XP_004843136.1 | 7 - 8 | 1147 | 83 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

LOC101696584| Gene Symbol: LOC101696584 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004843079.2 | XP_004843136.1 | 7 - 8 | 1147 | 83 | |
Set Membership: |
|