Assay Details
Gene Symbol: LOC101726067 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_004855792.3 | XP_004855849.1 | 11 - 12 | 1245 | 67 | |
XM_004855793.3 | XP_004855850.1 | 11 - 12 | 1246 | 67 | ||
XM_021237241.1 | XP_021092900.1 | 11 - 12 | 1246 | 67 |

More Information
Set Membership: |
![]() ![]() ![]() |
