Assay Details
Gene Symbol: LOC101726067 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_004855792.3 | XP_004855849.1 | 14 - 15 | 1689 | 109 | |
XM_004855793.3 | XP_004855850.1 | 14 - 15 | 1690 | 109 | ||
XM_021237241.1 | XP_021092900.1 | 14 - 15 | 1690 | 109 |

More Information
Set Membership: |
![]() ![]() ![]() |
