Assay Details
| Gene Symbol: Znf581 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004866626.3 | XP_004866683.1 | 1 - 2 | 168 | 111 | |
More Information
Additional Information:
This assay is in the 5'UTR region of the XM_004866626.3 transcripts.
Set Membership: |
|

Znf581
Genomic Map