Assay Details
| Gene Symbol: Rnf175 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004869208.3 | XP_004869265.1 | 7 - 8 | 960 | 61 | |
| XM_013067232.2 | XP_012922686.1 | 7 - 8 | 988 | 61 | ||
| XM_013067233.2 | XP_012922687.1 | 7 - 8 | 964 | 61 | ||
| XM_013067234.2 | XP_012922688.1 | 7 - 8 | 1088 | 61 | ||
| XM_021247874.1 | XP_021103533.1 | 7 - 8 | 1397 | 61 | ||
More Information
Set Membership: |
|

Rnf175
Genomic Map