Assay Details
Gene Symbol: Nacc1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_004872427.2 | XP_004872484.1 | 3 - 4 | 1167 | 67 | |
XM_021250350.1 | XP_021106009.1 | 4 - 5 | 1369 | 67 | ||
XM_021250351.1 | XP_021106010.1 | 4 - 5 | 1319 | 67 |

More Information
Set Membership: |
![]() ![]() ![]() |
