Assay Details
| Gene Symbol: Tmem255b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_021239364.1 | XP_021095023.1 | 5 - 6 | 461 | 61 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

Tmem255b| Gene Symbol: Tmem255b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_021239364.1 | XP_021095023.1 | 5 - 6 | 461 | 61 | |
Set Membership: |
|