Assay Details
Gene Symbol: LOC101697420 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_021240765.1 | XP_021096424.1 | 12 - 13 | 1259 | 82 | |
XM_021240766.1 | XP_021096425.1 | 12 - 13 | 1237 | 82 | ||
XM_021240767.1 | XP_021096426.1 | 12 - 13 | 1312 | 82 | ||
XM_021240768.1 | XP_021096427.1 | 12 - 13 | 1281 | 82 | ||
XM_021240769.1 | XP_021096428.1 | 11 - 12 | 1136 | 82 |

More Information
Set Membership: |
![]() ![]() ![]() |
Panther Classification:
