Assay Details
Gene Symbol: LOC101697420 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_021240765.1 | XP_021096424.1 | 5 - 6 | 615 | 87 | |
XM_021240766.1 | XP_021096425.1 | 5 - 6 | 593 | 87 | ||
XM_021240767.1 | XP_021096426.1 | 5 - 6 | 668 | 87 | ||
XM_021240768.1 | XP_021096427.1 | 5 - 6 | 637 | 87 | ||
XM_021240769.1 | XP_021096428.1 | 4 - 5 | 492 | 87 |

More Information
Set Membership: |
![]() ![]() |
Panther Classification:
